missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)benzonitrile, 98%, BLD Pharm™

Product Code. 30777189
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30777189 10 g 10g
30766875 25 g 25g
30766783 5 g 5g
30765195 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30777189 Supplier BLD Pharm Supplier No. BD1089510g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1483-54-1
Molecular Formula C8H5F3N2
MDL Number MFCD04974124
Quantity 10 g
SMILES N#CC1=CC=C(C(F)(F)F)C=C1N
Molecular Weight (g/mol) 186.13
Percent Purity 98%
Chemical Name or Material 2-Amino-4-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.