missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-1,3,4-thiadiazole, 97%, Thermo Scientific Chemicals

Product Code. 15362032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15362032 10 g 10g
15372032 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15362032 Supplier Thermo Scientific Acros Supplier No. 448670100
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37566-39-5
Molecular Formula C2H2BrN3S
Molecular Weight (g/mol) 180.02
MDL Number MFCD00464325
InChI Key GLYQQFBHCFPEEU-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-1,3,4-thiadiazole, 5-bromo-1,3,4-thiadiazol-2-ylamine, 2-amino-5-bromo-1,3,4 thiadiazole, 1,3,4-thiadiazol-2-amine, 5-bromo, 5-bromo-1,3,4-thiadiazole-2-amine, 5-bromo-1,3,4-thiadiazole-2-ylamine, 5-bromo-1,3,4 thiadiazol-2-ylamine, bromothiadiazolamine, pubchem20972, ksc496o1t
PubChem CID 3650883
IUPAC Name 5-bromo-1,3,4-thiadiazol-2-amine
SMILES NC1=NN=C(Br)S1

Specifications

Melting Point 167.0°C to 169.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 10 g
Formula Weight 180.03
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-1,3,4-thiadiazole
compliance-icons
Product Identifier
  • 2-Amino-5-bromo-1,3,4-thiadiazole , 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.