missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-4-methylpyridine, 97%

Product Code. 11435430
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435430 25 g 25g
11415430 1 g 1g
11425430 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435430 Supplier Thermo Scientific Alfa Aesar Supplier No. 044623.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98198-48-2 | C6H7BrN2 | 187.04 g/mol

2-Amino-5-bromo-4-methylpyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98198-48-2
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD03427660
InChI Key JDNCMHOKWINDKI-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-4-methylpyridine, 2-amino-4-methyl-5-bromopyridine, 2-amino-5-bromo-4-picoline, 5-bromo-4-methyl-pyridin-2-ylamine, 5-bromo-4-methyl-2-pyridinamine, 5-bromo-4-methyl-2-pyridinylamine, 2-pyridinamine, 5-bromo-4-methyl, 5-bromo-4-methyl-2-pyridylamine, 2-amino-5-bromo-4-picoline 2-amino-5-bromo-4-methylpyridine, pubchem2241
PubChem CID 817695
IUPAC Name 5-bromo-4-methylpyridin-2-amine
SMILES CC1=CC(=NC=C1Br)N

Specifications

Melting Point 148°C to 151°C
Color Cream
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 187.04
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-bromo-4-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.