missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-N-cyclohexyl-N-methylbenzylamine Dihydrochloride, TRC

Product Code. 30325828
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325828 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325828 Supplier Toronto Research Chemicals Supplier No. TRCA60192525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride
CAS 10076-98-9
Purity Grade Notes HPLC
Molecular Formula C14 H21 Br N2 . 2 Cl H
Quantity 25 mg
Synonym N-(2-Amino-5-bromobenzyl)-N-methyl-cyclohexanamine Dihydrochloride, Bromhexine Hydrochloride Imp. D (EP) as Dihydrochloride
InChI Formula InChI=1S/C14H21BrN2.2ClH/c1-17(13-5-3-2-4-6-13)10-11-9-12(15)7-8-14(11)16;;/h7-9,13H,2-6,10,16H2,1H3;2*1H
SMILES Cl.Cl.CN(Cc1cc(Br)ccc1N)C2CCCCC2
IUPAC Name 4-bromo-2-[[cyclohexyl(methyl)amino]methyl]aniline;dihydrochloride
Molecular Weight (g/mol) 370.16
Formula Weight 368.0422
Percent Purity >95
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.