missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzenesulfonic acid, 97%

Product Code. 11349949
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349949 5 g 5g
11359949 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349949 Supplier Thermo Scientific Alfa Aesar Supplier No. B25738.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 133-74-4 | C6H6ClNO3S | 207.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-74-4
Molecular Formula C6H6ClNO3S
Molecular Weight (g/mol) 207.63
MDL Number MFCD00035776
InChI Key ZCGVPUAAMCMLTM-UHFFFAOYSA-N
Synonym 2-amino-5-chlorobenzenesulphonic acid, 5-chloroorthanilic acid, 4-chloroaniline-2-sulfonic acid, 4-chloro-2-sulphoaniline, p-chloranilin-o-sulfosaure, p-chloroaniline-2-sulphonic acid, 2-amino-5-chloro-benzenesulfonic acid, benzenesulfonic acid,2-amino-5-chloro, 2-amino-5-chlorobenzene-1-sulfonic acid, 2-azanyl-5-chloranyl-benzenesulfonic acid
PubChem CID 8628
IUPAC Name 2-amino-5-chlorobenzenesulfonic acid
SMILES NC1=CC=C(Cl)C=C1S(O)(=O)=O

Specifications

Melting Point >300°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 207.64
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chlorobenzenesulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.