missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzonitrile, 97%, Thermo Scientific™

Product Code. 10070431 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10070431

Brand: Thermo Scientific Acros 326420050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 94.0°C to 99.0°C
CAS 5922-60-1
CAS Min % 96.0
Color White to Beige
CAS Max % 100.0
Boiling Point 132.0°C to 135.0°C (0.5 mmHg)
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Crystalline Needles
Assay Percent Range 97%
Molecular Formula C7H5ClN2
MDL Number MFCD00017106
Synonym 5-chloroanthranilonitrile, benzonitrile, 2-amino-5-chloro, anthranilonitrile, 5-chloro, 2-amino-5-chloro-benzonitrile, 4-chloro-2-cyanoaniline, unii-gy0x0082qb, 2-amino-5-chloro benzonitrile, amino 2-5-chloro-benzonitrile, 2-amino-5-chlorobenzenecarbonitrile, acbn
InChI Key QYRDWARBHMCOAG-UHFFFAOYSA-N
SMILES NC1=CC=C(Cl)C=C1C#N
IUPAC Name 2-amino-5-chlorobenzonitrile
Molecular Weight (g/mol) 152.58
PubChem CID 80019
Formula Weight 152.58
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chlorobenzonitrile, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.