missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-chlorobenzonitrile, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-5-chlorobenzonitrile |
| CAS | 5922-60-1 |
| Molecular Formula | C7H5ClN2 |
| Quantity | 5 g |
| Synonym | 5-Chloroanthranilonitrile, 1-Cyano-2-amino-5-chlorobenzene, 2-Amino-5-chlorobenzenecarbonitrile, 2-Cyano-4-chloroaniline, 4-Chloro-2-cyanoaniline, 5-Chloro-2-aminobenzonitrile, 5-Chloroanthranilonitrile, 6-Amino-3-chlorobenzonitrile |
| InChI Formula | InChI=1S/C7H5ClN2/c8-6-1-2-7(10)5(3-6)4-9/h1-3H,10H2 |
| SMILES | Nc1ccc(Cl)cc1C#N |
| IUPAC Name | 2-amino-5-chlorobenzonitrile |
| Molecular Weight (g/mol) | 152.58 |
| Formula Weight | 152.0141 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction