missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylthiazole, 98+%

Product Code. 15478929
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478929 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15478929 Supplier Thermo Scientific Alfa Aesar Supplier No. L11039.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7305-71-7 | C4H6N2S | 114.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7305-71-7
Melting Point 95°C to 98°C
Odor Recognizable
Molecular Formula C4H6N2S
MDL Number MFCD00078317
Quantity 5 g
UN Number UN3077
Beilstein 109603
Synonym 2-amino-5-methylthiazole, 5-methylthiazol-2-amine, 2-thiazolamine, 5-methyl, 2-amino-5-methyl-thiazole, thiazole, 2-amino-5-methyl, 5-methyl-2-thiazolamine, 5-methyl-2-aminothiazole, unii-7rab78af2p, 2-amino-5-methyl thiazole, meloxicam related compound b
InChI Key GUABFMPMKJGSBQ-UHFFFAOYSA-N
SMILES CC1=CN=C(S1)N
IUPAC Name 5-methyl-1,3-thiazol-2-amine
Molecular Weight (g/mol) 114.166
PubChem CID 351770
Formula Weight 114.17
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-methylthiazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.