missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-n-propylsulfonylbenzimidazole, 98+%

Product Code. 11393197
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393197 100 mg 100mg
11383197 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393197 Supplier Thermo Scientific Alfa Aesar Supplier No. L13711.MC
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80983-34-2 | C10H13N3O2S | 239.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80983-34-2
Molecular Formula C10H13N3O2S
Molecular Weight (g/mol) 239.29
MDL Number MFCD01075656
InChI Key WTPBIYSMFKUQKY-UHFFFAOYSA-N
Synonym albendazole-2-aminosulfone, 2-amino-5-propylsulphonylbenzimidazole, 2-amino-5-n-propylsulfonylbenzimidazole, 1h-benzimidazol-2-amine, 5-propylsulfonyl, 5-propylsulfonyl-1h-benzo d imidazol-2-amine, 2-amino-5-n-propylsulphonylbenzimidazole, abz2nh2, 5-propylsulfonyl-1h-benzimidazol-2-amine, 5-propylsulphonyl-1h-benzimidazol-2-amine, 5-propane-1-sulfonyl-1h-1,3-benzodiazol-2-amine
PubChem CID 88125
ChEBI CHEBI:80621
IUPAC Name 6-propylsulfonyl-1H-benzimidazol-2-amine
SMILES CCCS(=O)(=O)C1=CC=C2N=C(N)NC2=C1

Specifications

Melting Point 220°C to 222°C
Quantity 100 mg
Formula Weight 239.3
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-n-propylsulfonylbenzimidazole

RUO – Research Use Only

Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.