missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-naphthol-7-sulfonic Acid, TRC

Product Code. 30314833
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314833 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314833 Supplier Toronto Research Chemicals Supplier No. TRCA61869325G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-5-naphthol-7-sulfonic Acid
CAS 87-02-5
Molecular Formula C10H9NO4S
Quantity 25 g
Synonym 7-Amino-4-hydroxy-2-naphthalenesulfonic acid, 1-Hydroxy-3-sulfo-6-aminonaphthalene, 1-Hydroxy-6-amino-3-naphthalenesulfonic acid, 2-Amino-5-hydroxy-7-naphthalenesulfonic acid, 2-Amino-5-naphthol-7-sulfonic acid, 3-Amino-6-sulfo-8-naphthol, 3-Amino-8-hydroxy-6-naphthalenesulfonic acid, 3-Amino-8-hydroxy-6-sulfonaphthalene, 3-Amino-8-naphthol-6-sulfonic acid, 6-Amino-1-hydroxy-3-naphthalenesulfonic acid, 6-Amino-1-hydroxy-3-sulfonaphthalene, 6-Amino-1-naphthol-3-sulfonic acid, 6-Amino-3-sulfo-1-naphthol, I acid, Isogamma acid, J acid, NSC 31510, NSC 7556, NSC 863, 1
InChI Formula InChI=1S/C10H9NO4S/c11-7-1-2-9-6(3-7)4-8(5-10(9)12)16(13,14)15/h1-5,12H,11H2,(H,13,14,15)
SMILES Nc1ccc2c(O)cc(cc2c1)S(=O)(=O)O
IUPAC Name 7-amino-4-hydroxynaphthalene-2-sulfonic acid
Molecular Weight (g/mol) 239.25
Formula Weight 239.0252
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.