missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrothiazole, 97%

Product Code. 10674632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10674632 100 g 100g
10341334 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10674632 Supplier Thermo Scientific Acros Supplier No. 158521000
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-66-4 | C3H3N3O2S | 145.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-66-4
Molecular Formula C3H3N3O2S
Molecular Weight (g/mol) 145.14
MDL Number MFCD00005326
InChI Key MIHADVKEHAFNPG-UHFFFAOYSA-N
Synonym 2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix
PubChem CID 8486
ChEBI CHEBI:82386
IUPAC Name 5-nitro-1,3-thiazol-2-amine
SMILES NC1=NC=C(S1)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 196°C to 204°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Merck Index 15, 453
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in dilute mineral acids,1g/100g alcohol,1g/250g ether,practically insoluble in chloroform
Formula Weight 145.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-nitrothiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-nitrothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.