missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromobenzothiazole, 98%

Product Code. 11395199
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395199 5 g 5g
11385199 1 g 1g
11305209 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11395199 Supplier Thermo Scientific Alfa Aesar Supplier No. A18554.06
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15864-32-1 | C7H5BrN2S | 229.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15864-32-1
Molecular Formula C7H5BrN2S
Molecular Weight (g/mol) 229.10
MDL Number MFCD00152229
InChI Key VZEBSJIOUMDNLY-UHFFFAOYSA-N
Synonym 2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304
PubChem CID 85149
IUPAC Name 6-bromo-1,3-benzothiazol-2-amine
SMILES NC1=NC2=CC=C(Br)C=C2S1

Specifications

Odor Odorless
Quantity 5 g
Formula Weight 229.1
Percent Purity 98%
Chemical Name or Material 2-Amino-6-bromobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.