missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-bromopyridine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-6-bromopyridine |
| CAS | 19798-81-3 |
| Molecular Formula | C5H5BrN2 |
| Quantity | 10 g |
| Synonym | 2-Bromo-6-aminopyridine, 6-Bromo-2-aminopyridine, 6-Bromo-2-pyridinamine, 6-Bromopyridin-2-ylamine, NSC 176170 |
| InChI Formula | InChI=1S/C5H5BrN2/c6-4-2-1-3-5(7)8-4/h1-3H,(H2,7,8) |
| SMILES | Nc1cccc(Br)n1 |
| IUPAC Name | 6-bromopyridin-2-amine |
| Molecular Weight (g/mol) | 173.01 |
| Formula Weight | 171.9636 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction