missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-(chloromethyl)-4(3H)-pteridinone (>90%), TRC

Product Code. 30328057
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30328057 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30328057 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA60287550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-(Chloromethyl)pterin
CAS 873397-19-4
Molecular Formula C7 H6 Cl N5 O
Quantity 50 mg
Synonym 4(3H)-Pteridinone, 2-amino-6-(chloromethyl)-, 4-Pteridinol, 2-amino-6-(chloromethyl)- (5CI), 2-Amino-6-(chloromethyl)-4(3H)-pteridinone, 6-(Chloromethyl)pterin
InChI Formula InChI=1S/C7H6ClN5O/c8-1-3-2-10-5-4(11-3)6(14)13-7(9)12-5/h2H,1H2,(H3,9,10,12,13,14)
SMILES NC1=NC(=O)c2nc(CCl)cnc2N1
IUPAC Name 2-amino-6-(chloromethyl)-1H-pteridin-4-one
Molecular Weight (g/mol) 211.61
Formula Weight 211.0261
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.