missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropurine, TRC

Product Code. 30180284
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180284 50 g 50g
30179868 1 g 1g
30179819 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180284 Supplier Toronto Research Chemicals Supplier No. TRCA60350550G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10310-21-1
Molecular Formula C5 H4 Cl N5
Molecular Weight (g/mol) 169.57
Synonym 9H-Purin-2-amine, 6-chloro-, 1H-Purin-2-amine, 6-chloro- (9CI), Purine, 2-amino-6-chloro- (6CI,7CI,8CI), 6-Chloro-9H-purin-2-amine, 2-Amino-6-chloro-9H-purine, 2-Amino-6-chloropurine, 6-Chloro-1H-purin-2-amine, 6-Chloro-2-aminopurine, 6-Chloro-2-purinamine, 6-Chloro-7H-purin-2-amine, 6-Chloro-7H-purin-2-ylamine, 6-Chloro-9H-purin-2-yl-2-amine, 6-Chloroguanine, NSC 29570
IUPAC Name 6-chloro-9H-purin-2-amine
SMILES Nc1nc(Cl)c2nc[nH]c2n1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H4ClN5/c6-3-2-4(9-1-8-2)11-5(7)10-3/h1H,(H3,7,8,9,10,11)
Formula Weight 169.0155
Chemical Name or Material 2-Amino-6-chloropurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.