missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropyrazine, 95%

Product Code. 15408528
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408528 25 g 25g
15498518 5 g 5g
15418528 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15408528 Supplier Thermo Scientific Alfa Aesar Supplier No. H61934.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 33332-28-4 | C4H4ClN3 | 129.547 g/mol

2-Amino-6-chloropyrazine is used in the preparation of A2B adenosine receptor antagonists and other biologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chloropyrazine is used in the preparation of A2B adenosine receptor antagonists and other biologically active compounds.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33332-28-4
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD00055024
InChI Key JTPXVCKCLBROOJ-UHFFFAOYSA-N
Synonym 2-amino-6-chloropyrazine, 2-chloro-6-aminopyrazine, 2-pyrazinamine, 6-chloro, pyrazinamine, 6-chloro, 6-chloro-pyrazin-2-ylamine, 6-chloro-2-pyrazinamine, 6-chloropyrazin-2-ylamine, 2-amino-6-chloro-pyrazine, 6-chloropyrazine-2-ylamine, 6-amino-2-chloropyrazine
PubChem CID 118458
IUPAC Name 6-chloropyrazin-2-amine
SMILES C1=C(N=C(C=N1)Cl)N

Specifications

Quantity 25 g
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 129.55
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-6-chloropyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.