missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-mercaptopurine-13C2,15N, TRC

Product Code. 30012085
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012085 10 mg 10mg
30006233 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012085 Supplier Toronto Research Chemicals Supplier No. TRCA61183210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330266-29-9
Molecular Formula C313C2H5N415NS
Molecular Weight (g/mol) 170.17
Synonym 2-Amino-1,7(9)-dihydro-purine-6-thione-13C2,15N, Tioguanine-13C2,15N, 6-Thioguanine-13C2,15N, 2-Amino-9H-purine-6-thiol-13C2,15N;
IUPAC Name 2-amino-3,7-dihydropurine-6-thione
SMILES SC1=NC(N)=N[13C]2=[13C]1[15N]=CN2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C5H5N5S/c6-5-9-3-2(4(11)10-5)7-1-8-3/h1H,(H4,6,7,8,9,10,11)
Formula Weight 170.0303
Percent Purity >95%
Chemical Name or Material 2-Amino-6-mercaptopurine-13C2,15N
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.