missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-mercaptopurine, TRC

Product Code. 30137292
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137292 500 mg 500mg
18071340 250 mg 250mg
18071820 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137292 Supplier Toronto Research Chemicals Supplier No. TRCA611830500MG
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-42-7
Molecular Formula C5 H5 N5 S
Molecular Weight (g/mol) 167.19
Synonym 6H-Purine-6-thione, 2-amino-1,9-dihydro-, 6H-Purine-6-thione, 2-amino-1,7-dihydro- (9CI), Purine-6(1H)-thione, 2,3-dihydro-2-imino- (6CI), Purine-6(1H)-thione, 2-amino- (7CI,8CI), Purine-6-thiol, 2-amino- (8CI), 2-Amino-3,7-dihydropurine-6-thione, 2-Amino-6-mercaptopurine, 2-Amino-9H-purine-6(1H)-thione, 2-Amino-9H-purine-6-thiol, 2-Aminopurine-6-thiol, 6-Mercaptoguanine, 6-TG, 6-Thioguanine, Guanine, thio-, Lanvis, NSC 752, NSC 76504, Tabloid, Thioguanine, Thioguanine Tabloid, Tioguanin, Tioguanine
IUPAC Name 2-amino-1,7-dihydropurine-6-thione
SMILES NC1=Nc2nc[nH]c2C(=S)N1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H5N5S/c6-5-9-3-2(4(11)10-5)7-1-8-3/h1H,(H4,6,7,8,9,10,11)
Formula Weight 167.0266
Percent Purity >95%
Chemical Name or Material Thioguanine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.