missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxypyridine, 97%

Product Code. 15482749
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482749 5 g 5g
15492749 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482749 Supplier Thermo Scientific Alfa Aesar Supplier No. H66436.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 17920-35-3 | C6H8N2O | 124.143 g/mol

It's an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It′s an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17920-35-3
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD04972542
InChI Key DEUALFRBMNMGDS-UHFFFAOYSA-N
Synonym 2-amino-6-methoxypyridine, 6-methoxy-pyridin-2-ylamine, 6-methoxy-2-pyridinamine, 6-methoxypyrid-2-ylamine, 6-methoxy-2-pyridylamine, 2-amino-6-methoxy pyridine, 2-pyridinamine, 6-methoxy, methoxypyridin-6-amine, acmc-209efc, 6-methoxy-2-aminopyridin
PubChem CID 580053
IUPAC Name 6-methoxypyridin-2-amine
SMILES COC1=CC=CC(=N1)N

Specifications

Color Brown
Quantity 5 g
UN Number UN2810
Beilstein 115179
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 124.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Amino-6-methoxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.