missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylpyridine, 99%

Product Code. 11438663
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438663 500 g 500g
11428663 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438663 Supplier Thermo Scientific Alfa Aesar Supplier No. A10444.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1824-81-3 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1824-81-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006331
InChI Key QUXLCYFNVNNRBE-UHFFFAOYSA-N
Synonym 2-amino-6-methylpyridine, 2-amino-6-picoline, 6-amino-2-picoline, 2-pyridinamine, 6-methyl, 6-methyl-2-pyridinamine, 6-methyl-2-pyridylamine, 6-methyl-2-aminopyridine, 2-picoline, 6-amino, 6-amino-2-methylpyridine, pyridine, 2-amino-6-methyl
PubChem CID 15765
IUPAC Name 6-methylpyridin-2-amine
SMILES CC1=CC=CC(N)=N1

Specifications

Melting Point 41°C to 45°C
Boiling Point 208°C to 209°C
Flash Point 103°C (217°F)
Odor Unpleasant
Quantity 500 g
UN Number UN2811
Beilstein 107048
Sensitivity Hygroscopic
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2-Amino-6-methylpyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.