missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-9H-pyrido[2,3-b]indole, TRC

Product Code. 18092700
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18092700 5 mg 5mg
18070600 10 mg 10mg
18051610 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18092700 Supplier Toronto Research Chemicals Supplier No. TRCA6290005MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26148-68-5
Molecular Formula C11 H9 N3
Molecular Weight (g/mol) 183.21
Synonym 1H-Pyrido[2,3-b]indol-2-amine (9CI), 9H-Pyrido[2,3-b]indole, 2-amino- (8CI), 2-Amino-9H-pyrido[2,3-b]indole, 2-Amino-α-carboline, 9H-1,9-Diazafluoren-2-amine, AaC, AαC
IUPAC Name 9H-pyrido[2,3-b]indol-2-amine
SMILES Nc1ccc2c3ccccc3[nH]c2n1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H9N3/c12-10-6-5-8-7-3-1-2-4-9(7)13-11(8)14-10/h1-6H,(H3,12,13,14)
Formula Weight 183.0796
Percent Purity >95%
Chemical Name or Material 2-Amino-9H-pyrido[2,3-b]indole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.