missing translation for 'onlineSavingsMsg'
Learn More

2'-Aminoacetophenone, 97%

Product Code. 10627454 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627454 5 g Glass bottle 5g
10433351 10 g Glass bottle 10g
10208053 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10627454 Supplier Thermo Scientific Acros Supplier No. 400620050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 551-93-9 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 551-93-9
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007717
InChI Key GTDQGKWDWVUKTI-UHFFFAOYSA-N
Synonym 2'-aminoacetophenone, 1-2-aminophenyl ethanone, o-aminoacetophenone, 2-acetylaniline, o-acetylaniline, o-aminoacetylbenzene, 1-acetyl-2-aminobenzene, 1-2-aminophenyl ethan-1-one, acetophenone, 2'-amino, ethanone, 1-2-aminophenyl
PubChem CID 11086
IUPAC Name 1-(2-aminophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 20°C
Color Brown-Yellow to Yellow
Boiling Point 70°C to 71°C (3.0 mmHg)
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4COCH3
Refractive Index 1.6130 to 1.617
Quantity 5 g
Beilstein 14, 41
Merck Index 14, 413
Solubility Information Solubility in water: practically insoluble
Formula Weight 135.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2'-Aminoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2'-Aminoacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.