missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoanthracene, TRC

Product Code. 30324390
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324390 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324390 Supplier Toronto Research Chemicals Supplier No. TRCA57689050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 2-Aminoanthracene
CAS 613-13-8
Molecular Formula C14 H11 N
Quantity 50 mg
Synonym 2-Anthracenamine (9CI, ACI), 2-Anthramine (6CI, 7CI, 8CI), (Anthracen-2-yl)amine, 2-Aminoanthracene, 2-Anthracylamine, 2-Anthrylamine, NSC 400535, β-Aminoanthracene
InChI Formula InChI=1S/C14H11N/c15-14-6-5-12-7-10-3-1-2-4-11(10)8-13(12)9-14/h1-9H,15H2
SMILES Nc1ccc2cc3ccccc3cc2c1
IUPAC Name anthracen-2-amine
Molecular Weight (g/mol) 193.24
Formula Weight 193.0891
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.