missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminobenzene-1,4-disulfonic Acid (>90%), TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 2-Aminobenzene-1,4-disulfonic Acid |
| CAS | 98-44-2 |
| Molecular Formula | C6H7NO6S2 |
| Quantity | 500 mg |
| Synonym | 2-Amino-1,4-benzenedisulfonic Acid, 1-Amino-2,5-benzenedisulfonic Acid, 1-Anilino-2,5-disulfonic Acid, 2,5-Disulfo-1-aminobenzene, 2,5-Disulfoaniline, 2-Amino-1,4-disulfobenzene, 2-Amino-p-benzenedisulfonic Acid, 4-Sulfometanilic Acid, 6-Aminobenzene-1,4-disulfonic Acid, Aniline-2,5-disulfonic Acid, NSC 5580 |
| InChI Formula | InChI=1S/C6H7NO6S2/c7-5-3-4(14(8,9)10)1-2-6(5)15(11,12)13/h1-3H,7H2,(H,8,9,10)(H,11,12,13) |
| SMILES | Nc1cc(ccc1S(=O)(=O)O)S(=O)(=O)O |
| IUPAC Name | 2-aminobenzene-1,4-disulfonic acid |
| Molecular Weight (g/mol) | 253.24 |
| Formula Weight | 252.9715 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction