missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzeneboronic acid pinacol ester, 97%

Product Code. 11374316
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374316 5 g 5g
11364316 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374316 Supplier Thermo Scientific Alfa Aesar Supplier No. L19951.06
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 191171-55-8 | C12H18BNO2 | 219.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191171-55-8
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD02179448
InChI Key ZCJRWQDZPIIYLM-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-aminophenylboronic acid pinacol ester, 2-aminophenylboronic acid, pinacol ester, 2-aminophenylboronicacidpinacolester, 2-aminophenylboronic acid pinacol cyclic ester, 2-aminobenzeneboronic acid pinacol ester, 2-aminophenyl boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 2734652
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1N

Specifications

Melting Point 65°C to 70°C
Quantity 5 g
Beilstein 8548809
Sensitivity Air sensitive
Formula Weight 219.09
Percent Purity 97%
Chemical Name or Material 2-Aminobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.