missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Aminobutane, 98%

Product Code. 11459923
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
Product Code. Quantity unitSize
11459923 2500 mL 2500mL
11449923 500 mL 500mL
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 11459923

Marque: Thermo Scientific Alfa Aesar A13014.0F

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 13952-84-6 | C4H11N | 73.14 g/mol

(±)-2-Aminobutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Aminobutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive air and light. Incompatible with strong oxidizing agents. Incompatible with strong oxidizing agents, strong acids and carbon dioxide.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 13952-84-6
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00008094
InChI Key BHRZNVHARXXAHW-UHFFFAOYNA-N
Synonym sec-butylamine, 2-butanamine, 2-aminobutane, 2-butylamine, 1-methylpropylamine, butafume, tutane, 1-methylpropanamine, deccotane, frucote
PubChem CID 24874
ChEBI CHEBI:74526
IUPAC Name butan-2-amine
SMILES CCC(C)N

Spécification

Melting Point -104°C
Density 0.722
pH 12.5
Boiling Point 63°C to 64°C
Flash Point −19°C (−2°F)
Odor Amine-like
Refractive Index 1.393
Quantity 2500 mL
UN Number UN2733
Beilstein 1361345
Sensitivity Air Sensitive
Merck Index 14,1544
Solubility Information Soluble in water.
Formula Weight 73.14
Percent Purity 98%
Chemical Name or Material (+/-)-2-Aminobutane
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.