missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanethiol, 95%

Product Code. 11955751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11955751 25 g Glass bottle 25g
11331143 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11955751 Supplier Thermo Scientific Acros Supplier No. 434710250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-23-1 | C2H7NS | 77.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-23-1
Molecular Formula C2H7NS
Molecular Weight (g/mol) 77.15
MDL Number MFCD00008196
InChI Key UFULAYFCSOUIOV-UHFFFAOYSA-N
Synonym cysteamine, mercaptamine, thioethanolamine, becaptan, mercamine, cysteinamine, beta-mercaptoethylamine, 2-mercaptoethylamine, cysteamin, lambraten
PubChem CID 6058
ChEBI CHEBI:17141
IUPAC Name 2-aminoethanethiol
SMILES NCCS

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 95.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Iodimetry)
Packaging Glass bottle
Linear Formula NH2CH2CH2SH
Quantity 25 g
Merck Index 15,2776
Solubility Information Solubility in water: freely soluble.
Formula Weight 77.15
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Aminoethanethiol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminoethanethiol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.