missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl Diphenylborinate, TRC

Product Code. 30012118
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012118 1 g 1g
30010624 5 g 5g
30011621 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012118 Supplier Toronto Research Chemicals Supplier No. TRCA6091151G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-95-8
Molecular Formula C14H16BNO
Molecular Weight (g/mol) 225.09
Synonym Diphenylborinic Acid 2-Aminoethyl Ester, 2-Aminoethanol Diphenylborinate (Ester), (2-Aminoethyloxy)diphenylborane, 2-APB, 2-Aminoethoxydiphenyl Borate, 2-Aminoethoxydiphenylborane, 2-Aminoethyl Diphenylborinate, B-(2- Aminoethoxy)diphenylborane, NSC 17107, Naturstoffreagenz A
IUPAC Name 2-diphenylboranyloxyethanamine
SMILES NCCOB(c1ccccc1)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H16BNO/c16-11-12-17-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10H,11-12,16H2
Formula Weight 225.1325
Chemical Name or Material 2-Aminoethyl Diphenylborinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.