missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl hydrogen sulfate, 99%

Product Code. 10375780 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10375780

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 926-39-6 | C2H7NO4S | 141.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-39-6
Molecular Formula C2H7NO4S
Molecular Weight (g/mol) 141.14
MDL Number MFCD00008179
InChI Key WSYUEVRAMDSJKL-UHFFFAOYSA-N
Synonym ethanolamine o-sulfate, aminoethyl sulfate, 2-aminoethyl sulfate, mono 2-aminoethyl sulfate, 2-aminoethyl hydrogen sulphate, sulfuric acid mono 2-aminoethyl ester, was-34, 2-aminoethylhydrogensulfate, unii-9c8f910hlk, ethanol, 2-amino-, hydrogen sulfate ester
PubChem CID 70223
IUPAC Name 2-aminoethyl hydrogen sulfate
SMILES NCCOS(O)(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 277°C to 280°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NCH2CH2OSO3H
Quantity 100 g
Beilstein 04, 276
Solubility Information Solubility in water: soluble. Other solubilities: insolube in ethanol
Formula Weight 141.14
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Aminoethyl hydrogen sulfate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.