missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethyl methacrylate hydrochloride, 90%, stabilized

Product Code. 10680222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10680222 5 g Glass bottle 5g
10522612 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10680222 Supplier Thermo Scientific Acros Supplier No. 357810050
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2420-94-2 | C6H11NO2·HCl | 165.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2420-94-2
Molecular Formula C6H11NO2·HCl
Molecular Weight (g/mol) 165.62
MDL Number MFCD00078260
InChI Key XSHISXQEKIKSGC-UHFFFAOYSA-N
Synonym 2-aminoethyl methacrylate hydrochloride, 2-aminoethylmethacrylate hydrochloride, 2-aminoethyl 2-methylacrylate hydrochloride, methacrylic acid, 2-aminoethyl ester, hydrochloride, polymers, 2-aminoethyl 2-methylprop-2-enoate hydrochloride, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride 1:1, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride, aminoethylmethacrylate, acmc-209wgm, timtec-bb sbb003905
PubChem CID 75495
IUPAC Name 2-aminoethyl 2-methylprop-2-enoate;hydrochloride
SMILES CC(=C)C(=O)OCCN.Cl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 116.0°C to 118.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 88% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
Stabilizer 500 to 1000ppm phenothiazine
Formula Weight 165.62
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethyl methacrylate hydrochloride, 90%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminoethyl methacrylate hydrochloride, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.