missing translation for 'onlineSavingsMsg'
Learn More

(2-Aminoethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Product Code. 10722681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10722681 1 g Glass bottle 1g
10549073 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10722681 Supplier Thermo Scientific Acros Supplier No. 291630010
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2041-14-7
Molecular Formula C2H8NO3P
Molecular Weight (g/mol) 125.06
MDL Number MFCD00008182
InChI Key QQVDJLLNRSOCEL-UHFFFAOYSA-N
Synonym 2-aminoethyl phosphonic acid, ciliatine, phosphonoethylamine, aminoethylphosphonic acid, 2-aminoethane phosphonic acid, 2-aminoethanephosphonic acid, 2-aminoethylphosphonate, beta-aminoethylphosphonic acid, 2-aminoethyl phosphonate, phosphonic acid, 2-aminoethyl
PubChem CID 339
ChEBI CHEBI:15573
IUPAC Name 2-aminoethylphosphonic acid
SMILES NCCP(O)(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 296.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NH2CH2CH2P(O)(OH)2
Quantity 1 g
Beilstein 04, 1792
Solubility Information Solubility in water: soluble in water. Other solubilities: insoluble in methanol and acetone
Formula Weight 125.06
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (2-Aminoethyl)phosphonic acid
Show More Show Less
compliance-icons
Product Identifier
  • (2-Aminoethyl)phosphonic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.