missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoimidazole sulfate, 98%

Product Code. 11381899
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381899 10 g 10g
11371899 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381899 Supplier Thermo Scientific Alfa Aesar Supplier No. H25999.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1450-93-7 | C6H12N6O4S | 264.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1450-93-7
Molecular Formula C6H12N6O4S
Molecular Weight (g/mol) 264.26
MDL Number MFCD00013162
InChI Key KUWRLKJYNASPQZ-UHFFFAOYSA-N
Synonym 2-aminoimidazole hemisulfate, 1h-imidazol-2-amine sulfate 2:1, bis 1h-imidazol-2-amine ; sulfuric acid, 2-aminoimidazole sulphate, bis 2-aminoimidazole sulfate, 1h-imidazol-2-amine sulfurate, 2-aminoimidazole sulfate 2:1, 1h-imidazol-2-amine, sulfate 2:1, pubchem8378, aminoimidazole hemisulfate
PubChem CID 2734684
IUPAC Name 1H-imidazol-2-amine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=NC=CN1.NC1=NC=CN1

Specifications

Melting Point ∼270°C (decomposition)
Odor Odorless
Quantity 10 g
Beilstein 4726750
Formula Weight 132.13
Percent Purity 98%
Chemical Name or Material 2-Aminoimidazole sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.