missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric Acid (2-Methylalanine), TRC

Product Code. 30483419
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483419 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483419 Supplier Toronto Research Chemicals Supplier No. TRCM27657050G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminoisobutyric Acid (2-Methylalanine)
CAS 62-57-7
Molecular Formula C4H9NO2
Quantity 50 g
Synonym 2,2-Dimethylglycine, 2-Amino-2-methylpropanoic Acid, 2-Amino-2-methylpropionic Acid, 2-Methylalanine, 21: PN: WO2005055994 PAGE: 50 claimed sequence, AIB, Aminoisobutyric Acid, NSC 16590, alpha,alpha-Dimethylglycine, alpha-Aminoisobutanoic Acid, alpha-Aminoisobutyric Acid, alpha-Methylalanine
InChI Formula InChI=1S/C4H9NO2/c1-4(2,5)3(6)7/h5H2,1-2H3,(H,6,7)
SMILES CC(C)(N)C(=O)O
IUPAC Name 2-amino-2-methylpropanoic acid
Molecular Weight (g/mol) 103.12
Formula Weight 103.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.