missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric acid, 98+%

Product Code. 10388740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10388740 25 g 25g
10124900 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10388740 Supplier Thermo Scientific Acros Supplier No. 225760250
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 62-57-7 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-57-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00008049
InChI Key FUOOLUPWFVMBKG-UHFFFAOYSA-N
Synonym 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid
PubChem CID 6119
ChEBI CHEBI:27971
IUPAC Name 2-amino-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 335.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CH3)2C(NH2)CO2H
Quantity 25 g
Beilstein 04, 414
Merck Index 15, 442
Solubility Information Solubility in water: soluble. Other solubilities: difficultly soluble in alcohol, insoluble in ether
Formula Weight 103.12
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoisobutyric acid, ≥98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.