missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoisobutyric acid, 99%

Product Code. 11489933
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489933 250 g 250g
11389679 25 g 25g
11399679 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489933 Supplier Thermo Scientific Alfa Aesar Supplier No. A13021.30
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 62-57-7 | C4H9NO2 | 103.121 g/mol

2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoisobutyric acid is a non- protein amino acid present in some antibiotics. It acts as a strong helix inducer in peptides. It is non-metabolisable used in bioassays.

Solubility
Soluble in water. Insoluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-57-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008049
InChI Key FUOOLUPWFVMBKG-UHFFFAOYSA-N
Synonym 2-aminoisobutyric acid, 2-methylalanine, alanine, 2-methyl, alpha-aminoisobutyric acid, 2-amino-2-methylpropionic acid, h-aib-oh, alpha-methylalanine, alpha-aminoisobutanoic acid, a-aminoisobutyric acid, 2-amino isobutyric acid
PubChem CID 6119
ChEBI CHEBI:27971
IUPAC Name 2-amino-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)N

Specifications

Melting Point >300°C
Quantity 250 g
Beilstein 506496
Merck Index 14,445
Solubility Information Soluble in water. Insoluble in alcohol and ether.
Formula Weight 103.12
Percent Purity 99%
Chemical Name or Material 2-Aminoisobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.