missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminoisobutyric acid methyl ester hydrochloride, 99%
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Descripción
Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system and in studies on amino acid transport systems.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Especificaciones
Especificaciones
| Melting Point | ∽185°C (decomposition) |
| Quantity | 1 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 153.61 |
| Percent Purity | 99% |
| Chemical Name or Material | 2-Aminoisobutyric acid methyl ester hydrochloride |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido