missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)benzimidazole dihydrochloride hydrate, 98%

Product Code. 11331917
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331917 25 g 25g
11321917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331917 Supplier Thermo Scientific Alfa Aesar Supplier No. L07440.14
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5993-91-9 | C8H11Cl2N3 | 220.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5993-91-9
Molecular Formula C8H11Cl2N3
Molecular Weight (g/mol) 220.10
MDL Number MFCD00012677
InChI Key HAEYZZSSSUIZAN-UHFFFAOYSA-N
Synonym 1h-benzo d imidazol-2-yl methanamine dihydrochloride, 2-aminomethyl benzimidazole dihydrochloride, 1h-benzimidazole-2-methanamine, dihydrochloride, 1h-1,3-benzodiazol-2-ylmethanamine dihydrochloride, 1h-1,3-benzodiazol-2-yl methanamine dihydrochloride, 2-aminomethylbenzimidazole dihydrochloride, c-1h-benzoimidazol-2-yl-methylamine dihydrochloride, benzimidazol-2-ylmethylamine, chloride, chloride, 1-1h-1,3-benzodiazol-2-yl methanamine dihydrochloride, 1h-benzo d imidazol-2-yl methanamine 2hcl
PubChem CID 2723957
IUPAC Name 1H-benzimidazol-2-ylmethanamine;dihydrochloride
SMILES Cl.Cl.NCC1=NC2=CC=CC=C2N1

Specifications

Melting Point ∽264°C (decomposition)
Quantity 25 g
Beilstein 3698942
Formula Weight 220.10 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 2-(Aminomethyl)benzimidazole dihydrochloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.