missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)indole, 97%

Product Code. 15490598
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490598 250 mg 250mg
15470598 1 g 1g
15480598 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490598 Supplier Thermo Scientific Alfa Aesar Supplier No. H54281.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21109-25-1 | C9H10N2 | 146.193 g/mol

2-(Aminomethyl)indole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Aminomethyl)indole is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21109-25-1
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD03422512
InChI Key RNAODKZCUVVPEN-UHFFFAOYSA-N
Synonym 1h-indol-2-ylmethyl amine, 1h-indol-2-yl methanamine, 1h-indole-2-methanamine, 1-1h-indol-2-yl methanamine, 2-aminomethyl indole, 2-aminomethylindole, c-1h-indol-2-yl-methylamine, 2-indolemethylamine, 1h-indol-2-yl methylamine, indol-2-ylmethylamine
PubChem CID 582331
IUPAC Name 1H-indol-2-ylmethanamine
SMILES C1=CC=C2C(=C1)C=C(N2)CN

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 2-(Aminomethyl)indole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.