missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)piperidine, 98%

Product Code. 11354566
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354566 5 g 5g
11364566 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354566 Supplier Thermo Scientific Alfa Aesar Supplier No. L20122.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22990-77-8 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22990-77-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00129011
InChI Key RHPBLLCTOLJFPH-UHFFFAOYNA-N
Synonym 2-piperidylmethylamine, 2-aminomethyl piperidine, 2-piperidinemethanamine, 2-aminomethylpiperidine, 2-aminomethyl-1-piperidine, 1-piperidin-2-yl methanamine, piperidine-2-methylamine, piperidine, 2-aminomethyl, pubchem6725, 2-aminomethyl-piperdine
PubChem CID 90865
IUPAC Name piperidin-2-ylmethanamine
SMILES NCC1CCCCN1

Specifications

Density 0.942
Boiling Point 63°C to 66°C (12 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.485
Quantity 5 g
UN Number UN2735
Beilstein 102994
Sensitivity Air Sensitive
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 2-(Aminomethyl)piperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.