missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)pyridine, 98+%

Product Code. 11460807
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460807 100 g 100g
11440807 5 g 5g
11450807 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460807 Supplier Thermo Scientific Alfa Aesar Supplier No. B23208.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3731-51-9 | C6H8N2 | 108.14 g/mol

2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.

Solubility
Soluble in water (1000 mg/ml at 20°C), and ethanol.

Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, acids, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3731-51-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006360
InChI Key WOXFMYVTSLAQMO-UHFFFAOYSA-N
Synonym 2-picolylamine, 2-aminomethyl pyridine, 2-pyridinemethanamine, 2-aminomethylpyridine, 2-pyridinemethylamine, 2-pyridylmethylamine, 2-picolinamine, 2-pyridinemethaneamine, 2-pyridylmethyl amine, pyridine, 2-aminomethyl
PubChem CID 19509
ChEBI CHEBI:81387
SMILES NCC1=CC=CC=N1

Specifications

Melting Point -20°C
Density 1.049
pH 11 to 12
Boiling Point 202°C to 204°C
Flash Point 90°C (194°F)
Odor Amine-like
Refractive Index 1.545
Quantity 100 g
UN Number UN2735
Beilstein 108054
Sensitivity Air Sensitive
Solubility Information Soluble in water (1000mg/ml at 20°C),and ethanol.
IUPAC Name pyridin-2-ylmethanamine
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2-(Aminomethyl)pyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.