missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)pyridine, 99%

Product Code. 11956281 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11956281 500 mL Glass bottle 500mL
10589610 25 mL Glass bottle 25mL
10142750 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11956281 Supplier Thermo Scientific Acros Supplier No. 104075000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3731-51-9 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3731-51-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006360
InChI Key WOXFMYVTSLAQMO-UHFFFAOYSA-N
Synonym 2-picolylamine, 2-aminomethyl pyridine, 2-pyridinemethanamine, 2-aminomethylpyridine, 2-pyridinemethylamine, 2-pyridylmethylamine, 2-picolinamine, 2-pyridinemethaneamine, 2-pyridylmethyl amine, pyridine, 2-aminomethyl
PubChem CID 19509
ChEBI CHEBI:81387
IUPAC Name pyridin-2-ylmethanamine
SMILES NCC1=CC=CC=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -20.0°C
Color Colorless to Yellow or Orange
Density 1.0400g/mL
Boiling Point 82.0°C to 85.0°C (12.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5420 to 1.5460
Quantity 500 mL
Specific Gravity 1.04
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 108.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Aminomethyl)pyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Aminomethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.