missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenylboronic acid hydrochloride, 97%

Product Code. 11996121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11996121 1 g 1g
11906131 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11996121 Supplier Thermo Scientific Acros Supplier No. 445310010
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 863753-30-4 | C6H9BClNO2 | 173.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 863753-30-4
Molecular Formula C6H9BClNO2
Molecular Weight (g/mol) 173.40
MDL Number MFCD02258096
InChI Key WPDASZCYRKGSTO-UHFFFAOYSA-N
Synonym 2-aminophenylboronic acid hydrochloride, 2-aminophenyl boronic acid hydrochloride, 2-aminophenylboronic acid hcl, 2-aminophenylboronic acid, hydrochloride, 2-2-aminophenyl boronic acid hydrochloride, boronic acid, 2-aminophenyl-, hydrochloride, pubchem1746, ksc914i9f, 2-aminophenylboronic acid,hcl, 2-aminophenylboronic acid, hcl
PubChem CID 16427090
SMILES Cl.NC1=CC=CC=C1B(O)O

Specifications

Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name (2-aminophenyl)boronic acid hydrochloride
Formula Weight 173.41
Percent Purity 97%
Chemical Name or Material 2-Aminophenylboronic acid hydrochloride
compliance-icons
Product Identifier
  • 2-Aminophenylboronic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.