missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine, 99%

Product Code. 11499513
Click to view available options
Quantity:
25 g
250 g
1000 g
Unit Size:
1000g
250g
25g
This item is not returnable. View return policy

Product Code. 11499513

Brand: Thermo Scientific Alfa Aesar A12374.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 504-29-0 | C5H6N2 | 94.117 g/mol

A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.

Solubility
Slightly soluble in water(8g/100 mL at 19°C)

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-29-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006312
InChI Key ICSNLGPSRYBMBD-UHFFFAOYSA-N
Synonym 2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino
PubChem CID 10439
IUPAC Name pyridin-2-amine
SMILES C1=CC=NC(=C1)N

Specifications

Melting Point 56°C to 60°C
Boiling Point 209°C to 211°C
Flash Point 92°C (197°F)
Quantity 1000 g
UN Number UN2671
Beilstein 105785
Sensitivity Hygroscopic
Merck Index 14,473
Solubility Information Slightly soluble in water(8g/100 mL at 19°C)
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material 2-Aminopyridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.