missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyrimidine, 98%

Product Code. 10794011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10794011 500 g Plastic bottle 500g
10793821 5 g Glass bottle 5g
10234320 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10794011 Supplier Thermo Scientific Acros Supplier No. 104585000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-12-6 | C4H5N3 | 95.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-12-6
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00006089
InChI Key LJXQPZWIHJMPQQ-UHFFFAOYSA-N
Synonym 2-aminopyrimidine, 2-pyrimidinamine, pyrimidin-2-ylamine, pyrimidinamine, aminopyrimidine, 2-pyrimidiylamine, pyrimidine, 2-amino, 2-pyridiylamine, 1,2-dihydro-2-iminopyrimidine, pyrimidine, amino
PubChem CID 7978
ChEBI CHEBI:38618
IUPAC Name pyrimidin-2-amine
SMILES NC1=NC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 124°C to 127°C
Color White to Yellow
Boiling Point 158°C (186.0 Mbar)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 500 g
Beilstein 24, 80
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetic acid,readily soluble in alcohol,acetone and chloroform,slightly soluble in ether and benzene
Formula Weight 95.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Aminopyrimidine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.