missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole-5-carboxaldehyde, 95%

Product Code. 15413617
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15413617 Supplier Thermo Scientific Alfa Aesar Supplier No. H27112.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1003-61-8 | C4H4N2OS | 128.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1003-61-8
Melting Point ∽160°C (decomposition)
Molecular Formula C4H4N2OS
MDL Number MFCD02179570
Quantity 1 g
Sensitivity Air Sensitive
Synonym 2-amino-5-formylthiazole, 2-aminothiazole-5-carbaldehyde, 2-aminothiazole-5-carboxaldehyde, 2-amino-thiazole-5-carbaldehyde, 2-amino-5-formyl-1,3-thiazole, 5-thiazolecarboxaldehyde, 2-amino, 2-amino-1,3-thiazole-5-carboxaldehyde, zlchem 77, pubchem8904, 5-formyl-2-aminothiazole
InChI Key HZKMBJCDAXLMDN-UHFFFAOYSA-N
SMILES C1=C(SC(=N1)N)C=O
IUPAC Name 2-amino-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol) 128.149
PubChem CID 2737798
Formula Weight 128.15
Percent Purity 95%
Chemical Name or Material 2-Aminothiazole-5-carboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.