missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiobenzamide, 97%

Product Code. 15488267
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488267 5 g 5g
15478267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488267 Supplier Thermo Scientific Alfa Aesar Supplier No. H51809.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2454-39-9 | C7H8N2S | 152.215 g/mol

It can be heated to produce 2-trifluoromethyl-3H-quinazoline-4-thione in the presence of reagent molecular sieves and solvent CHCl3 with reaction time of 2 hours. It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be heated to produce 2-trifluoromethyl-3H-quinazoline-4-thione in the presence of reagent molecular sieves and solvent CHCl3 with reaction time of 2 hours. It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2454-39-9
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.215
MDL Number MFCD00963496
InChI Key HPZKAJRFABCGFF-UHFFFAOYSA-N
Synonym 2-aminothiobenzamide, 2-amino-thiobenzamide, 2-aminobenzothioamide, benzenecarbothioamide, 2-amino, thioanthranilamide, o-aminothiobenzamide, 2-aminobenzene-1-carbothioamide, amino 2-aminophenyl methane-1-thione, benzamide, o-aminothio, thioanthranilsaureamid
PubChem CID 1550990
IUPAC Name 2-aminobenzenecarbothioamide
SMILES C1=CC=C(C(=C1)C(=S)N)N

Specifications

Melting Point 115°C to 120°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 152.21
Percent Purity 97%
Chemical Name or Material 2-Aminothiobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.