missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiophenol 90%, TRC

Product Code. 30309007
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309007 Supplier Toronto Research Chemicals Supplier No. TRCA63042525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminothiophenol
CAS 137-07-5
Molecular Formula C6H7NS
Quantity 25 g
Synonym o-Aminobenzenethiol, 1-Amino-2-mercaptobenzene, 2-Amino-1-mercaptobenzene, 2-Aminobenzenethiol, 2-Aminophenyl Mercaptan, 2-Aminothiophenol, 2-Mercaptoaniline, 2-Thioaniline, NSC 106635, o-Aminobenzenethiol, o-Aminophenyl Mercaptan, o-Aminothiophenol, o-Mercaptoaniline, alpha-Aminothiophenol
InChI Formula InChI=1S/C6H7NS/c7-5-3-1-2-4-6(5)8/h1-4,8H,7H2
SMILES Nc1ccccc1S
IUPAC Name 2-aminobenzenethiol
Molecular Weight (g/mol) 125.19
Formula Weight 125.0299
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.