missing translation for 'onlineSavingsMsg'
Learn More

(2-anilinopyrimidin-5-yl)methylamine, 95%, Thermo Scientific™

Product Code. 10603085
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10603085 250 mg Amber Glass Bottle 250mg
10613085 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10603085

Brand: Thermo Scientific Maybridge CC69213CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1093860-47-9
Molecular Formula C11H12N4
Molecular Weight (g/mol) 200.245
MDL Number MFCD11841074
InChI Key IGMKIKUXMJLMOP-UHFFFAOYSA-N
Synonym 5-aminomethyl-n-phenylpyrimidin-2-amine, 2-anilinopyrimidin-5-yl methylamine, 5-aminomethyl-2-phenylamino pyrimidine, 5-aminomethyl pyrimidin-2-yl phenylamine
PubChem CID 43811051
IUPAC Name 5-(aminomethyl)-N-phenylpyrimidin-2-amine
SMILES C1=CC=C(C=C1)NC2=NC=C(C=N2)CN

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 200.24
Percent Purity 95%
Chemical Name or Material (2-Anilinopyrimidin-5-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.