missing translation for 'onlineSavingsMsg'
Learn More

2-Anilinopyrimidine-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10194194
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10194194 1 g Amber Glass Bottle 1g
10092074 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10194194 Supplier Thermo Scientific Maybridge Supplier No. CC69201DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 450368-25-9
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.21
MDL Number MFCD09864415
InChI Key ZPIYMRAJXNMUPE-UHFFFAOYSA-N
Synonym 2-phenylamino pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-phenylamino, 2-anilino-5-pyrimidinecarboxylic acid, 2-anilinopyrimidine-5-carboxylic acid
PubChem CID 23509722
IUPAC Name 2-anilinopyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=C(NC2=CC=CC=C2)N=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.21
Percent Purity 97%
Chemical Name or Material 2-Anilinopyrimidine-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.