missing translation for 'onlineSavingsMsg'
Learn More

(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%

Product Code. 10414035 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10414035 1 g 1g
10053480 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10414035 Supplier Thermo Scientific Acros Supplier No. 297060010
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 49805-30-3 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49805-30-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00213364
InChI Key DDUFYKNOXPZZIW-UHFFFAOYSA-N
Synonym 2-azabicyclo 2.2.1 hept-5-en-3-one, +/--2-azabicyclo 2.2.1 hept-5-en-3-one, 3-azabicyclo 2.2.1 hept-5-en-2-one, inverted exclamation marka-2-azabicyclo 2.2.1 hept-5-en-3-one, vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, --2-azabicyclo 2.2.1 hept-5-en-3-one, +-2-azabicyclo 2.2.1 hept-5-en-3-one
PubChem CID 548678
IUPAC Name 3-azabicyclo[2.2.1]hept-5-en-2-one
SMILES C1C2C=CC1NC2=O

Specifications

Absorbance (c=1, CH2Cl2) (1cm cell), (c=1, CH2Cl2) (1cm cell), 2 max. at 400nm
CAS Min % 97.5
CAS Max % 100.0
Melting Point 55.0°C to 59.0°C
Color White to Beige
Boiling Point 102°C to 106°C (0.2 hPa)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: >1000g/L (23°C).
Formula Weight 109.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (#1)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.